Learn More
Tocris Bioscience™ TSA Vivid Fluorophore kit 520
Click to view available options
Quantity:
1 Kit (20 to 80 Slides)
Unit Size:
1 set
Description
TSA Vivid reagents are exceptionally bright dyes that offer an effective way to boost signal intensity and detect low-abundance targets in spatial biology applications. They are designed for use in ISH, IHC and ICC experiments where in situ detection of target protein or nucleic acid sequences is required. TSA Vivid reagents have been specifically optimized to deliver exceptional signal-to-noise in the RNAScope™ Multiplex Fluorescent v2 Assay for visualization of gene expression at the single cell level. They are suitable for multiplexing and can be combined with DAPI (Cat. No. 5748) counter-staining.
The basis for signal amplification comes from the specific and high-density deposition of TSA Vivid dyes adjacent to a horseradish peroxidase (HRP)-labeled probe in a process termed Tyramide Signal Amplification (TSA) or Catalyzed Reporter Deposition (CARD).
Excitation maximum = 494 nm; emission maximum = 517 nm
Please see the product datasheet or protocol for information on product use.
Specifications
Specifications
| Quantity | 1 Kit (20 to 80 Slides) |
| Target | Tyramide Signaling Amplification/TSA |