Learn More
TSEN34 Monoclonal Antibody (OTI6B3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810146
Description
TSEN34 encodes a catalytic subunit of the tRNA splicing endonuclease, which catalyzes the removal of introns from precursor tRNAs. The endonuclease complex is also associated with a pre-mRNA 3-prime end processing factor. A mutation in this gene results in the neurological disorder pontocerebellar hypoplasia type 2. Multiple alternatively spliced variants, encoding the same protein, have been identified.Specifications
| TSEN34 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BSV6 | |
| TSEN34 | |
| Full length human recombit protein of human TSEN34 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6B3 | |
| Unconjugated | |
| TSEN34 | |
| LENG5, PCH2C, SEN34, SEN34L | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 79042 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.