Learn More
TSEN34 Monoclonal Antibody (OTI6B3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810146S
Description
TSEN34 Monoclonal antibody specifically detects TSEN34 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| TSEN34 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y3S2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 27309 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6B3 | |
| Unconjugated | |
| ZNF330 | |
| CTD-3093M3.1; hsSen34; LENG5; leukocyte receptor cluster (LRC) member 5; leukocyte receptor cluster member 5; PCH2C; SEN34; SEN34L; tRNA splicing endonuclease subunit 34; tRNA-intron endonuclease Sen34; tRNA-splicing endonuclease subunit Sen34; TSEN34; TSEN34 tRNA splicing endonuclease subunit | |
| Full length human recombinant protein of human ZNF330 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.