missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSLP Antibody (55N1E3), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27323B
This item is not returnable.
View return policy
Description
TSLP Monoclonal specifically detects TSLP in Human samples. It is validated for Western Blot.
Specifications
| TSLP | |
| Monoclonal | |
| Biotin | |
| Q969D9 | |
| TSLP | |
| amino acids (27-158) MDYDFTNCDFEkIkAAYLSTISkDLITYMSGTkSTEFNNTVSCSNRPHCLTEIQSLTFNP∼TAGCASLAkEMFAMkTkAALAIWCPGYSETQINATQAMkkRRkRkVTTNkCLEQVSQLQG∼LWRRFNRPLLkQQL | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 55N1E3 | |
| Western Blot | |
| thymic stromal lymphopoietin | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 85480 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction