missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN7/TM4SF2 Antibody (CL0265) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44062
This item is not returnable.
View return policy
Description
TSPAN7/TM4SF2 Monoclonal antibody specifically detects TSPAN7/TM4SF2 in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunohistochemistry
Specifications
| TSPAN7/TM4SF2 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| A15CCG-B7, CD231, CD231 antigen, Cell surface glycoprotein A15, Membrane component chromosome X surface marker 1, mental retardation, X-linked 58, MRX58, MXS1T-cell acute lymphoblastic leukemia associated antigen 1, T-cell acute lymphoblastic leukemia-associated antigen 1, tetraspanin 7, tetraspanin-7, TM4SF2tetraspanin protein, transmembrane 4 superfamily 2b, Transmembrane 4 superfamily member 2X chromosome, surface marker 1, transmembrane protein A15, Tspan-7 | |
| This antibody was generated using a recombinant protein sequence of P41732, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| metabolism | |
| 7102 | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin), Immunohistochemistry | |
| CL0265 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction