Learn More
TST Antibody (5E11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43056
This item is not returnable.
View return policy
Description
TST Monoclonal antibody specifically detects TST in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| TST | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot | |
| 500000000000 | |
| Western Blot 1:500 - 1:5000, ELISA, Immunohistochemistry 1:20 - 1:200 | |
| EC 2.8.1, EC 2.8.1.1, MGC19578, RDSRhodanese, thiosulfate sulfurtransferase, thiosulfate sulfurtransferase (rhodanese) | |
| thiosulfate sulfurtransferase(rhodanese) - (Uniprot# Q16762) | |
| 100 μg | |
| Proteases & Other Enzymes | |
| 7263 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |