missing translation for 'onlineSavingsMsg'
Learn More

TSTU, TRC

Product Code. 30454500
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454500 1 g -
30451093 5 g 5g
30450617 50 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30454500 Supplier Toronto Research Chemicals Supplier No. TRCT8955601G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105832-38-0
Molecular Formula C9 H16 N3 O3 . B F4
Molecular Weight (g/mol) 301.046
Synonym O-(N-Succinimidyl)-1,1,3,3-tetramethyluronium Tetrafluoroborate, N-[(Dimethylamino)[(2,5-dioxo-1-pyrrolidinyl)oxy]methylene]-N-methylmethanaminium Tetrafluoroborate(1-), N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium Tetrafluoroborate, O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate
SMILES [F-][B+3]([F-])([F-])[F-].CN(C)C(=[N+](C)C)ON1C(=O)CCC1=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H16N3O3.BF4/c1-10(2)9(11(3)4)15-12-7(13)5-6-8(12)14;2-1(3,4)5/h5-6H2,1-4H3;/q+1;-1
Formula Weight 301.122
Chemical Name or Material TSTU
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.