missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00€ - 572.00€
Specifications
| Antigen | TTC12 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18261232
|
Novus Biologicals
NBP2-58627 |
100 μL |
572.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627986
|
Novus Biologicals
NBP2-58627-25ul |
25 μL |
415.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TTC12 Polyclonal specifically detects TTC12 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TTC12 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| FLJ13859, FLJ20535, tetratricopeptide repeat domain 12, tetratricopeptide repeat protein 12 variant 1, tetratricopeptide repeat protein 12 variant 2, TPARMtetratricopeptide repeat protein 12, TPR repeat protein 12 | |
| TTC12 | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 54970 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FLDFSDKEANTAMGLFTDLALEERFQVWFQANLPGVLPALTGVLKTDPKVSSSSALCQCIAIMGNLSAEPTTRRHMAACEEFGDGC | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title