Learn More
TTC23 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand:  Novus Biologicals H00064927-B01P
Product Code. 18483779
  • 589.00€ / 50µg
Description
TTC23 Polyclonal antibody specifically detects TTC23 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TTC23
Polyclonal
Western Blot 1:500, Immunocytochemistry/ Immunofluorescence
AAH15728.1
Mouse
Protein A purified
RUO
64927
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunocytochemistry/Immunofluorescence
Unconjugated
PBS (pH 7.4)
Cervical cancer proto-oncogene 8 protein, FLJ12939, HCC-8FLJ12572, proto-oncogene 8, tetratricopeptide repeat domain 23, tetratricopeptide repeat protein 23, TPR repeat protein 23
TTC23 (AAH15728.1, 1 a.a. - 393 a.a.) full-length human protein. MQESQETHISNHLDEVVAAVSITHRKKFQNKLLQTALFQPPREKLHLCEEKAKSYSNSHEYKQAVHELVRCVALTRICYGDSHWKLAEAHVNLAQGYLQLKGLSLQAKQHAEKARQILANSIVPPYSENTDVFKFSIELFHTMGRALLSLQKFKEAAENLTKAERLSKELLQCGRIIKEEWIEIEARIRLSFAQVYQGQKKSKEALSHYQAALEYVEISKGETSRECVPILRELAGVEQALGLHDVSINHFLQAHLIILSRSPSQVEAADSAHIVAHAAVASGRHEHHDVAEQYFQESMAHLKDSEGMGRTKFLSIQDEFCHFLQMTGQKERATSILRESLEAKVEAFGDFSPEVAETYRLLGGADLAQGNHSGARKKLKKGDLSFLLSATLP
0.05 mg