Learn More
TTF1 Monoclonal Antibody (OTI7E8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
242.00€ - 540.87€
Specifications
| Antigen | TTF1 |
|---|---|
| Clone | OTI7E8 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17508224
|
Origene Technologies
TA813358S |
30 μL |
242.00€
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17588214
|
Origene Technologies
TA813358 |
100 μL |
540.87€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
TTF1 Monoclonal antibody specifically detects TTF1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| TTF1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| AV245725; BCH; BHC; Homeobox protein NK-2 homolog A; homeobox protein Nkx-2.1; NK-2; NK2 homeobox 1; NK-2 homolog A; NKX2.1; NKX2-1; NKX2A; NMTC1; RGD1565673; RNA polymerase I termination factor; RNA polymerase I transcription termination factor; T/EBP; TEBP; Thyroid nuclear factor 1; thyroid transcription factor 1; thyroid transcription factor 1-like; thyroid-specific enhancer-binding protein; TITF1; Transcription termination factor 1; transcription termination factor I; transcription termination factor, RNA polymerase I; TTF1; TTF-1; TTF-I | |
| Full length human recombinant protein of human TTF1 (NP_003308) produced in E.coli. | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Ttf1 |
| OTI7E8 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| Q15361, Q62187 | |
| 22130, 499766, 7270 | |
| IgG1 | |
| Protein A/G | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.