Learn More
TTPA Mouse anti-Human, Unconjugated, Clone: 3E7D10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67431-1-IG-20UL
Description
The TTPA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TTPA. This antibody recognizes human, mouse, rat, pig antigen. The TTPA antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| TTPA | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 3E7D10 | |
| Unconjugated | |
| 20 μL | |
| 7274 | |
| IgG2b |