Learn More
BD Vacutainer™ Luer Adapters
Product Code. 15120716
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Unit Size
15120716 Sterile Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15120716 Supplier BD Supplier No. 367300
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ideal for use with any female luer fitting. BD Vacutainer™ Luer Adapters are sterile devices with a multi-sampling valve that are designed for use with a catheter or hypodermic needle to collect blood using BD Vacutainer™ blood collection tubes.
The BD Vacutainer™ system is a closed system optimally designed and tested for controlling pre-analytical variability throughout the specimen collection and handling process.
  • Provides sterile, secure and safe specimen sampling for fast, accurate and consistent results
  • Offers the security of a threaded, locking luer connection, replacing a luer-slip device
  • Latex-free and compatible with a female luer connection or needleless IV site designed for luer lock access
  • Used together with the BD Vacutainer™ single-use tube holder and evacuated blood collection tubes
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
For Use With (Application) Blood collection from a catheter, needleless IV site, hypodermic needle or any female luer fitting; For safe and easy subculturing
For Use With (Equipment) Vacutainer™ Single-use Tube Holders and Vacutainer™ Evacuated Blood Collection Tubes
Includes With luer adapter
Latex Free Yes
Product Type Tube Adapters and Holder
Sterility Sterile