Learn More
Löser™ Drainage Tube for Osmometer
Product Code. 11960557
Change view
Click to view available options
For Use With (Equipment):
Osmometer Type 15/16
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11960557 Osmometer Type 15/16 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11960557 Supplier Löser™ Supplier No. 0.01.0007
  • ONLINE EXCLUSIVE PRICE
    2.20€
    1.68€ / Pack of 1
    Save 0.52€
    23% Off
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Provides precision while cleaning the needle and needle hole of an automatic osmometer. Loeser™ Drainage Tube for Osmometer fits into to Drainage Bottle.
  • Used with Drainage Bottle
  • For cleaning needle and needle hole of automatic osmometers
  • Length: 300mm
Specifications
Product Type Drainage Tube 300 mm
For Use With (Equipment) Osmometer Type 15/16
For Use With (Application) Cleaning osmometer needle and needle hole