missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FITC Monoclonal Antibody (NAWESLEE), Biotin, eBioscience™

FITC Monoclonal Antibody (NAWESLEE), Biotin, eBioscience™
Brand: Affymetrix eBioscience 13-7691-82
This item is not returnable.
View return policy
Description
Specifications
| FITC | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid | |
| Chemical |
| Flow Cytometry | |
| NAWESLEE | |
| Biotin | |
| Fluorescein | |
| Affinity Chromatography | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction