Learn More
TUBG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA589901
Description
Immunogen sequence: PWGPASIQVA LSRKSPYLPS AHRVSGLMMA NHTSISSLFE RTCRQYDKLR KREAFLEQFR KEDMFKDNFD EMDTSREIVQ QLIDEYHAAT RPDYISWGTQ E; Positive Samples: MCF-7, HeLa, Mouse brain, Mouse lung, Rat brain; Cellular Location: Cytoplasm, centrosome, cytoskeleton, microtubule organizing center
This gene encodes a member of the tubulin superfamily. The encoded protein localizes to the centrosome where it binds to microtubules as part of a complex referred to as the gamma-tubulin ring complex. The protein mediates microtubule nucleation and is required for microtubule formation and progression of the cell cycle. A pseudogene of this gene is found on chromosome 7.
Specifications
| TUBG1 | |
| Polyclonal | |
| Unconjugated | |
| TUBG1 | |
| 1500010O08Rik, AI451582, AI503389, CDCBM4, gamma-1-tubulin, Gamma-tubulin complex component 1, GCP-1, Tubg, TUBG1, TUBGCP1, tubulin gamma 1, tubulin gamma-1 chain, tubulin, gamma 1, tubulin, gamma polypeptide | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 103733, 252921, 7283 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 2.35 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal, pH 7.3 | |
| P23258, P83887, P83888 | |
| TUBG1 | |
| A synthetic peptide corresponding to a sequence within amino acids 350-450 of human γ-Tubulin (NP_0010612) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.