Learn More
Cole-Parmer Tubing Cutters
Product Code. 30053156 Shop All Cole-Parmer Products
Change view
Click to view available options
For Use With:
1/16 in. to 5/8 in. OD Tubing
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
30053156 1/16 in. to 5/8 in. OD Tubing Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30053156 Supplier Cole-Parmer Supplier No. 5011848
  • ONLINE EXCLUSIVE PRICE
    15.70€
    15.39€ / Pack of 1
    Save 0.31€
    1% Off
View all promotions
Estimated Shipment: 11-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Durable steel blade makes straight, clean cuts through plastic tubing,Blue.
Steel blade offers durability
Specifications
Material Steel
For Use With 1/16 in. to 5/8 in. OD Tubing
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.