missing translation for 'onlineSavingsMsg'
Learn More

Tulathromycin B, approximately 5% (As a mixture with Tulathromycin A), TRC

Product Code. 17933815
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17933815 50 mg 50mg
17944185 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17933815 Supplier Toronto Research Chemicals Supplier No. TRCT89715550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 217500-96-4∣280755-12-6
Molecular Formula C41 H79 N3 O12
Molecular Weight (g/mol) 806.08
Synonym (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-13-[[2,6-Dideoxy-3-C-methyl-3-O-methyl-4-C-[(propylamino)methyl]-α-L-ribo-hexopyranosyl]oxy]-2-ethyl-3,4,10-trihydroxy-3,5,8,10,12,14-hexamethyl-11-[[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylo-hexopyranosyl]oxy]-1-oxa-6-azacyclopentadecan-15-one, CP 472295, Draxxin, Tulathromycin, Tulathromycin A, (2R,3R,6R,8R,9R,10S,11S,12R)-11-[[2,6-dideoxy-3-C-methyl-3-O-methyl-4-C-[(propylamino)methyl]-alpha-L-ribo-hexopyranosyl]oxy]-2-[(1R,2R)-1,2-dihydroxy-1-methylbutyl]-8-hydroxy-3,6,8,10,12-pentamethyl-9-[[3,4,6-trideoxy-3-(dimethylamino)-beta-D-xylo-hexopyranosyl]oxy]-1-oxa-4-azacyclotridecan-13-one (9CI), CP 547272, Tulathromycin B
IUPAC Name (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-11-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-2-ethyl-3,4,10-trihydroxy-13-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyl-5-(propylaminomethyl)oxan-2-yl]oxy-3,5,8,10,12,14-hexamethyl-1-oxa-6-azacyclopentadecan-15-one∣(2S,3S,6R,8R,9R,10S,11S,12R)-2-[(2R,3R)-2,3-dihydroxypentan-2-yl]-9-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-8-hydroxy-11-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyl-5-(propylaminomethyl)oxan-2-yl]oxy-3,6,8,10,12-pentamethyl-1-oxa-4-azacyclotridecan-13-one
SMILES CCCNC[C@]1(O)[C@H](C)O[C@H](C[C@@]1(C)OC)O[C@H]2[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)CN[C@H](C)[C@@H](O)[C@](C)(O)[C@@H](CC)OC(=O)[C@@H]2C∣CCCNC[C@]1(O)[C@H](C)O[C@H](C[C@@]1(C)OC)O[C@H]2[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)CN[C@@H](C)[C@H](OC(=O)[C@@H]2C)[C@](C)(O)[C@H](O)CC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C41H79N3O12/c1-15-17-42-22-41(50)28(8)53-31(20-39(41,10)51-14)55-33-25(5)35(56-37-32(45)29(44(12)13)18-24(4)52-37)38(9,48)19-23(3)21-43-27(7)34(46)40(11,49)30(16-2)54-36(47)26(33)6/h23-35,37,42-43,45-46,48-50H,15-22H2,1-14H3/t23-,24-,25+,26-,27-,28+,29+,30-,31+,32-,33+,34-,35-,37+,38-,39-,40-,41+/m1/s1∣InChI=1S/C41H79N3O12/c1-15-17-42-22-41(50)28(8)53-31(20-39(41,10)51-14)54-33-25(5)34(56-37-32(46)29(44(12)13)18-24(4)52-37)38(9,48)19-23(3)21-43-27(7)35(55-36(47)26(33)6)40(11,49)30(45)16-2/h23-35,37,42-43,45-46,48-50H,15-22H2,1-14H3/t23-,24-,25+,26-,27+,28+,29+,30-,31+,32-,33+,34-,35+,37+,38-,39-,40-,41+/m1/s1
Formula Weight 805.5664
Percent Purity >95%
Chemical Name or Material Tulathromycin A∣Tulathromycin B
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.