missing translation for 'onlineSavingsMsg'
Learn More

Tulobuterol, TRC

Product Code. 17952695
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17952695 100 mg 100mg
17951915 50 mg 50mg
17964365 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17952695 Supplier Toronto Research Chemicals Supplier No. TRCT897250100MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41570-61-0
Molecular Formula C12 H18 Cl N O
Molecular Weight (g/mol) 227.73
Synonym Benzenemethanol, 2-chloro-α-[[(1,1-dimethylethyl)amino]methyl]-, 2-Chloro-α-[[(1,1-dimethylethyl)amino]methyl]benzenemethanol, (±)-Tulobuterol, HN 078, Tulobuterol, o-Chloro-α-[(tert-butylamino)methyl]benzyl alcohol
IUPAC Name 2-(tert-butylamino)-1-(2-chlorophenyl)ethanol
SMILES CC(C)(C)NCC(O)c1ccccc1Cl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H18ClNO/c1-12(2,3)14-8-11(15)9-6-4-5-7-10(9)13/h4-7,11,14-15H,8H2,1-3H3
Formula Weight 227.1077
Percent Purity >95%
Chemical Name or Material Tulobuterol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.