Learn More
Rogo Sampaic™ Tungsten Carbide Anatomical Heavy Duty Scissors
Product Code. 11736145
Change view
Click to view available options
Tip Style:
Round/Round
Sharp/Round
Sharp/Sharp
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tip Style Unit Size
11736145 Round/Round Each
11726145 Sharp/Sharp Each
11716145 Sharp/Round Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11736145 Supplier Rogo Sampaic™ Supplier No. FMN031
  • ONLINE EXCLUSIVE PRICE
    29.95€
    25.71€ / Pack of 1
    Save 4.24€
    14% Off
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Product Type Heavy Duty Anatomical Scissors
Length (Metric) 140 mm
Material Tungsten Carbide
Tip Style Round/Round