Learn More
Rogo Sampaic™ Tungsten Carbide Iris Scissor
Product Code. 11746145
Change view
Click to view available options
Length (Metric):
110 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
11746145 110 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11746145 Supplier Rogo Sampaic™ Supplier No. FMN023
  • ONLINE EXCLUSIVE PRICE
    29.45€
    25.28€ / Pack of 1
    Save 4.17€
    14% Off
In Stock
Add to basket
Add to basket
Specifications
Product Type Iris Scissors
Length (Metric) 110 mm
Material Tungsten Carbide
Tip Style Sharp/Sharp