missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TUP1 Monoclonal Antibody (10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA112683
This item is not returnable.
View return policy
Description
A suggested positive control for this product is Mav108 cells expressing recombinant TUP1 protein.
TUP 1 and SSN6 are involved in repression of several diverse families of genes in yeast, including cell type-specific genes regulated by the alphga2 and a1/alpha2 repressors. Ssn6-Tupl interacts with class I histone deacetylases required for repression, and rfecrfuitment of yeast Tuplp-Ssn6p repressor is associated with localized decreases in histone acetylation.
Specifications
| TUP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.08% sodium azide | |
| P16649 | |
| TUP1 | |
| Recombinant protein corresponding to the residues 1-200 of yeast TUP1 protein. | |
| 100 μg | |
| Primary | |
| Yeast | |
| Antibody | |
| IgG |
| Western Blot | |
| 10 | |
| Unconjugated | |
| TUP1 | |
| AAR1; AER2; AMM1; chromatin-silencing transcriptional regulator TUP1; CRT4; CYC9; FLK1; Flocculation suppressor protein; General transcriptional corepressor TUP1; Glucose repression regulatory protein TUP1; Repressor AER2; ROX4; SFL2; TUP1; UMR7; YCR084C; YCR84C | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 850445 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur