Learn More
Berthold Technologies Turn Table For DeWit Tubes
Product Code. 31777388 Shop All Berthold Technologies Products
Change view
Click to view available options
For Use With (Equipment):
Imaging System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
31777388 Imaging System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31777388 Supplier Berthold Technologies Supplier No. 5662503
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For NightSHADE imaging systems to hold 18 DeWit tubes for sequential imaging.
  • The turntable for NightSHADE enables sequential and scheduled acquisition of up to 18 DeWitt tubes (in groups of three)
  • Includes complete turntable assembly: base and upper part
Specifications
Type Specimen Tray
For Use With (Application) Imaging of Seedlings
For Use With (Equipment) Imaging System
Voltage 24 V