Learn More
TUSC3 Mouse anti-Human, Unconjugated, Clone: 1D3G12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67382-1-IG-20UL
Description
The TUSC3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TUSC3. This antibody recognizes human antigen. The TUSC3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TUSC3 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, ELISA | |
| 1D3G12 | |
| Unconjugated | |
| 20 μL | |
| 7991 | |
| IgG1 |