missing translation for 'onlineSavingsMsg'
Learn More

Tween-20 (Technical Grade), TRC

Product Code. 30006816
Change view
Click to view available options
Quantity:
50 mL
100 mL
Unit Size:
100mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006816 100 mL 100mL
30011022 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006816 Supplier Toronto Research Chemicals Supplier No. TRCT897285100ML
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-64-5
Synonym Ahco 7596T, Alkamuls PSML 20, Alkamuls PSML 80/72, Alkamuls T 20, Alkest TW 20, Anapoe 20, Armotan PML 20, Atlas G 4280, Atlas G 7596J, Atlas G 7596P, Atmer 110, Canarcel TW 20, Coulter Dispersant IB, Cremophor PS 20, Crillet 1, Crillet 1HP, D 941, Disponil SML 120, Disponil SML 20, E 432, Emasol 1112, Emasol L 120V, Emasol L 130, Emsorb 6915, Ercasorb 2020, Ethoxylated Sorbitan Monolaurate, Ethsorbox L 20, Ethylene Oxide-sorbitan Monolaurate Adduct, Ethylene Oxide-sorbitan Monolaurate Polymer, Eumulgin SML 15, Eumulgin SML 20, G 1020, G 4280, G 7596J, G 7606J, GL 1, Glycosperse L 20K, Glytanox 1001, Ionet T 20C, Kaopan TW-L 120, Kemotan T 20, Kolliphor PS 20, LT 221, Lamesorb SML 20, Liposorb L 20, Lumisorb PSML-80, ML 55F, Monopol Twl 1030, Montanox 20, Nikkol LT 221, Nikkol TL 10, Nissan Nonion LT 204, Nissan Nonion LT 210, Nissan Nonion LT 221, Nissan Nonion LT 280, Nonion LT 210, Nonion LT 221, Nonion LT 280, Oxyethylated Sorbitan Monolaurate, Oxysorbic 20, P 1397, PEG Sorbitan Monolaurate, PEG-20 Sorbitan Monolaurate, PEO(20) Sorbitan Monolaurate, POE Sorbitan Monolaurate, PTV-M 5309, Pionin D 941, Poly(ethylene glycol) Sorbitan Ether Monolaurate, Poly(oxyethylene sorbitan laurate), Poly(oxyethylene)sorbitan Ether Monolaurate, Polyethylene Glycol Sorbitan Monolaurate, Polyoxyethylene Span 20, Polyoxyethylene Sorbitan Monododecanoate, Polyoxyethylene Sorbitan Monolaurate, Polysorbate 20, Polysorbate 20PH, Polysorbate 21, Polysorbate E 432, Polysten 20, Protasorb L 20, Radiasurf 7137, Rheodol PW-S 106, Sorbimacrogol Laurate, Sorbimacrogol Laurate 300, Sorbitan Monolaurate Poly(oxyethylene)ether, Sorbitan Monolaurate Polyethylene Glycol Ether, Sorbitan Polyethoxy Monolaurate, Sorbon T 20, Sorboxethane Laurate, Sorgen TW 20, Sorgen TW 20F, T-MAZ 20, TW-L 120 Super, Tego SML 20, Tween 10, Tween 164, Tween 21, Tween 327, Tween L 120, Value 7220, Vykamol Sorbitol 6B, Wilsurf TF 20, Witconol 2720;
SMILES OCCO[C@@H]1[C@@H]([C@H](OCCO)COCCOC(CCCCCCCCCCC)=O)OC[C@H]1OCCO

Specifications

Quantity 100 mL
Chemical Name or Material Polysorbate 20
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.