Learn More
Sonics Materials™ Twenty-four-element Probe
Product Code. 11723589
Change view
Click to view available options
Packaging Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity Unit Size
11723589 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11723589 Supplier Sonics Materials™ Supplier No. 6300579
  • ONLINE EXCLUSIVE PRICE
    4000.00€
    3616.69€ / Pack of 1
    Save 383.31€
    9% Off
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Product Type 24-element Probe
Includes Aluminum Coupler and 24 x 3 mm Special Microtips
Packaging Quantity 1 Ea.