Description
- Reduction of consumable-related carbon footprint in the lab
- Laser-engraved lot number and expiration date on each single plate to improve traceability in your laboratory processes
- Unique laser-engraved optical guiding grid and OptiTrack matrix for quick orientation when pipetting manually
- One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
- Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
- Exceptionally solid, torque- and heat-resistant polycarbonate frame: No warping, bending or breaking during heat incubation or centrifugation
- Raised well rims for effective sealing, also reduces risk of cross-contamination
- Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
- Available with barcode (upon request)
Specifications
Specifications
| Barcode | Custom Barcoding Available |
| Format | SBS Standard |
| Array | 8 x 12 |
| Product Line | Eppendorf |
| Color | Clear |
| Product Type | BioBased PCR Plates |
| Skirt Style | Skirted |
| Material | Polypropylene, Polycarbonate |
| Volume (Metric) Well | 150 μL |
| No. of Wells | 96 |
| Show More |