missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TXNDC16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569614
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Dog: 86%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%.
TXNDC16 gene ontology annotations related to this gene include cell redox homeostasis.
Specifications
| TXNDC16 | |
| Polyclonal | |
| Unconjugated | |
| TXNDC16 | |
| 5730420B22Rik; C77647; ERp90; Kiaa1344; RGD1306755; thioredoxin domain containing 16; thioredoxin domain-containing protein 16; TXNDC16 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 57544 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9P2K2 | |
| TXNDC16 | |
| synthetic peptide directed towards the N terminal region of human TXNDC16. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction