Learn More
TXNDC5 Mouse, anti-Human, Clone: 1A12C3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68609-1-IG-150UL
 View more versions of this product
Product Code. 18058985
  • 410.00€ / 150µL
Description
The TXNDC5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TXNDC5. This antibody recognizes human, mouse antigen. The TXNDC5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
TXNDC5
1A12C3
EndoPDI, ER protein 46, ERP46, Hcc 2, PDIA15, Thioredoxin like protein p46, TLP46, TXNDC5, UNQ364
150 μL
81567
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human, Mouse
150 kDa