Learn More
TXNDC5 Mouse, anti-Human, Clone: 1A12C3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68609-1-IG-150UL
Description
The TXNDC5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TXNDC5. This antibody recognizes human, mouse antigen. The TXNDC5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TXNDC5 | |
| 1A12C3 | |
| EndoPDI, ER protein 46, ERP46, Hcc 2, PDIA15, Thioredoxin like protein p46, TLP46, TXNDC5, UNQ364 | |
| 150 μL | |
| 81567 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG1 |