Learn More
Saint Gobain Life Sciences Tygon™ E-3603 Tubing
Product Code. 15108106
Change view
Click to view available options
Diameter (Metric) Outer:
4 mm
8 mm
11.1 mm
22.3 mm
31.8 mm
Unit Size:
15m
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
15108106 8 mm 15m
15690211 4 mm 15m
10140121 22.3 mm 15m
15610221 11.1 mm 15m
15517995 31.8 mm 15m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15108106 Supplier Saint Gobain Life Sciences Supplier No. ACF00012S
  • ONLINE EXCLUSIVE PRICE
    122.48€
    81.27€ / 15m
    Save 41.21€
    33% Off
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Tygon™ E-3603 Tubing for laboratory and vacuum applications.
Tygon™ E-3603 tubing from Saint-Gobain is crystal clear and flexible, handling virtually all organic chemicals found in the modern laboratory. It is non-oxidizing and non-contaminating, making it an ideal choice for a variety of applications. Tygon™ E-3603 has high tolerances and lot-to-lot consistency, which will yield reproducible results in your laboratory. It is non-toxic and non-contaminating, and less permeable than rubber tubing. The glassy-smooth inner bore helps prevent buildup to facilitate cleaning.
  • Lot-to-lot consistency for reproducible results
  • Non-toxic and non-contaminating
  • Smooth, polished inner wall
  • Slips easily over fittings and grips securely for simple lab set-ups
  • Contains no BPA or phthalates
  • Standard sizes available to hold full vacuum at room temperature
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance FDA, NSF 51, EU Food, Japan Food, California Prop 65, USP Class VI
Autoclavable Autoclavable
Diameter (English) Inner 0.031 in.
Diameter (Metric) Inner 0.8 mm
Material PVC
Diameter (English) Outer 0.315 in.
Diameter (Metric) Outer 8 mm
Overall Length (English) 50 ft.
Overall Length (Metric) 15 m
Thickness (English) 0.0625 in.
Thickness (Metric) 1.5875 mm
For Use With Food and beverage dispensing
For Use With (Application) For condensers, incubators, desiccators, gas lines and drain lines
Min. Operating Temperature (English) -51°F
Min. Operating Temperature (Metric) -46°C
Packaging Quantity 1 Ea.
Resistance Properties 173 lb-f/in
Sterility Non-sterile
Color Clear
Hardness 56 (Shore A)
Flexibility 16 mm bend radius
Maximum Pressure 25 psi
Vacuum Level 29.9 in. Hg (at 73°F)
Max. Operating Temperature (English) 165°F
Max. Operating Temperature (Metric) 74°C
Product Type Tubing