missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tylosin tartrate 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Tylosin tartrate |
| CAS | 74610-55-2 |
| Analyte or Component Names | Tylosin tartrate |
| Shipping Condition | +20°C |
| Molecular Formula | C46H77NO17.C4H6O6 |
| Concentration | 1000 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Tylosin (2R,3R)-2,3-dihydroxybutanedioate (1:1) (salt), Tylosin [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1) (salt), Oxacyclohexadecane, tylosin deriv., Pharmasin, Tylan Injectable, Tylan Soluble, Tylosin tartrate, Tylosin Tartrate |
| InChI Formula | InChI=1S/C46H77NO17.C4H6O6/c1-13-33-30(22-58-45-42(57-12)41(56-11)37(52)26(5)60-45)18-23(2)14-15-31(49)24(3)19-29(16-17-48)39(25(4)32(50)20-34(51)62-33)64-44-38(53)36(47(9)10)40(27(6)61-44)63-35-21-46(8,55)43(54)28(7)59-35;5-1(3(7)8)2(6)4(9)10/h14-15,17-18,24-30,32-33,35-45,50,52-55H,13,16,19-22H2,1-12H3;1-2,5-6H,(H,7,8)(H,9,10)/b15-14+,23-18+;/t24-,25+,26-,27-,28+,29+,30-,32-,33-,35+,36-,37-,38-,39-,40-,41-,42-,43+,44-,45-,46-;1-,2-/m11/s1 |
| SMILES | CC[C@H]1OC(=O)C[C@@H](O)[C@H](C)[C@@H](O[C@H]2O[C@H](C)[C@@H](O[C@H]3C[C@@](C)(O)[C@@H](O)[C@H](C)O3)[C@@H]([C@H]2O)N(C)C)[C@@H](CC=O)C[C@@H](C)C(=O)\C=C\C(=C\[C@@H]1CO[C@@H]4O[C@H](C)[C@@H](O)[C@@H](OC)[C@H]4OC)\C.O[C@H]([C@@H](O)C(=O)O)C(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?