missing translation for 'onlineSavingsMsg'
Learn More

Tyloxapol, TRC

Product Code. 30449067
Change view
Click to view available options
Quantity:
1 g
2.5 g
25 g
Unit Size:
1g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449067 1 g 1g
30452133 25 g 25g
30454857 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30449067 Supplier Toronto Research Chemicals Supplier No. TRCT9477001G
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25301-02-4
Molecular Formula ((C H2 O)mon(C2 H4 O)mon(C14 H22 O)mon)co
Molecular Weight (g/mol) 280.4
Synonym p-(1,1,3,3-Tetramethylbutyl)Phenol Polymer with Ethylene Oxide and Formaldehyde, Ethylene Oxide Polymer with Formaldehyde and p-(1,1,3,3-Tetramethylbutyl)phenol, Oxirane Polymer with Formaldehyde and 4-(1,1,3,3-tetramethylbutyl)phenol, Ethylene oxide-formaldehyde-4-(1,1,3,3-tetramethylbutyl)phenol copolymer, Ethylene oxide-formaldehyde-p-octylphenol Copolymer, NSC 90255, Superinone, Triton A 20, Triton WR 1339, Tyloxapol, p-Isooctylpolyoxyethylenephenol Formaldehyde Polymer
IUPAC Name formaldehyde;oxirane;4-(1,1,3,3-tetramethylbutyl)phenol
SMILES CC(C)(C)CC(C)(C)c1ccc(O)cc1.C=O.C2CO2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C14H22O.C2H4O.CH2O/c1-13(2,3)10-14(4,5)11-6-8-12(15)9-7-11;1-2-3-1;1-2/h6-9,15H,10H2,1-5H3;1-2H2;1H2
Formula Weight 280.2038
Chemical Name or Material Tyloxapol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.