Learn More
TYRP1 Antibody (CL4906) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61142MFV450
Product Code. 30291384
  • 759.00€ / 0.10mL
Description
TYRP1 Monoclonal antibody specifically detects TYRP1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TYRP1
Monoclonal
mFluor Violet 450 SE
b-PROTEIN, CAS25,6-dihydroxyindole-2-carboxylic acid oxidase, Catalase B, CATB, EC 1.14.18, EC 1.14.18.-, EC 1.14.18.1, Glycoprotein 75, GP75DHICA oxidase, Melanoma antigen gp75, TRP-1, TRPTYRP, tyrosinase-related protein 1TRP1, TYRRP
This antibody was developed using a recombinant protein derived from P17643, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer
7306
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL4906