Learn More
DuPont™ Tyvek™ 500 Xpert BioCircle™ CHF5
GREENER_CHOICE
Product Code. 31903166 Shop All DuPont Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
31903166 Medium Pack of 25
31903158 Small Pack of 25
31903164 Large Pack of 25
31903156 X-Large Pack of 25
31903154 2X-Large Pack of 25
31903162 3X-Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31903166 Supplier DuPont™ Supplier No. D15582585
Robust yet lightweight protection providing a barrier against fine particles and low-level sprays in a hooded coverall using white Tyvek™ fabric attributed to 100% bio-circular HDPE through an ISCC-certified mass balance approach.
Features stitched external seams, a respirator fit three-piece hood, an ergonomic design, elasticated wrists, ankles, face, and waist (glued-in), and a Tyvek™ zipper with a pin lock slider zipper pull and a storm flap. Suitable for a variety of applications including pharmaceutical handling, chemical processing, general maintenance and operations, and automatic spray painting.
  • Independent cradle-to-gate life cycle analysis, including biogenic carbon, shows at least 58% lower climate change impact compared to the standard Tyvek™ 500 Xpert coverall
  • Certified according to Regulation (EU) 2016/425
  • Chemical protective clothing, Category III, Type 5-B and 6-B
  • EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination)
  • Antistatic treatment (EN 1149-5) - on both sides
  • Stitched external seams
  • Very low inward leakage thanks to optimised design
  • Suitable for use in GMP class C/D (ISO class 6-9) clean rooms
  • High liquid and particulate protection
  • Exceptional design and comfort
  • Good breathability thanks to air and moisture vapour permeability
  • Overall ergonomic shape for perfect fit and protection when moving
TRUSTED_SUSTAINABILITY
Specifications
Size Medium
Gender Unisex
Material HDPE
Color White
Disposable Single-use
Closure Type Zipper with Storm Flap
Sterility Non-sterile
Resistance Chemicals, Liquid, Particle, Anti-static, Dust
Latex Free Yes
No. of Pockets None