Description
Features stitched external seams, a respirator fit three-piece hood, an ergonomic design, elasticated wrists, ankles, face, and waist (glued-in), and a Tyvek™ zipper with a pin lock slider zipper pull and a storm flap. Suitable for a variety of applications including pharmaceutical handling, chemical processing, general maintenance and operations, and automatic spray painting.
- Independent cradle-to-gate life cycle analysis, including biogenic carbon, shows at least 58% lower climate change impact compared to the standard Tyvek™ 500 Xpert coverall
- Certified according to Regulation (EU) 2016/425
- Chemical protective clothing, Category III, Type 5-B and 6-B
- EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination)
- Antistatic treatment (EN 1149-5) - on both sides
- Stitched external seams
- Very low inward leakage thanks to optimised design
- Suitable for use in GMP class C/D (ISO class 6-9) clean rooms
- High liquid and particulate protection
- Exceptional design and comfort
- Good breathability thanks to air and moisture vapour permeability
- Overall ergonomic shape for perfect fit and protection when moving
Specifications
Specifications
| Size | Medium |
| Gender | Unisex |
| Material | HDPE |
| Color | White |
| Disposable | Single-use |
| Closure Type | Zipper with Storm Flap |
| Sterility | Non-sterile |
| Resistance | Chemicals, Liquid, Particle, Anti-static, Dust |
| Latex Free | Yes |
| No. of Pockets | None |