Learn More
DuPont™ Tyvek™ IsoClean Disposable Shoe Covers
Product Code. 15590611 Shop All DuPont Products
Change view
Click to view available options
Size:
Medium
Large
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15590611 Large Pack of 100
15500631 Medium Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15590611 Supplier DuPont™ Supplier No. D14246699
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
IsoClean overshoes are an ideal balance of protection, durability, and comfort.
  • Fixation ties
  • Elastics at ankles and gripper slip-retardant full sole
  • Tyvek™ garments and accessories are composed of flash spun high-density polythylene, providing an ideal balance of protection, durability and comfort
  • Tyvek™ is permeable to both air and water vapor, yet repels water-based liquids and aerosols
  • Garments and accessories made of Tyvek™ IsoClean are used in the biotech, pharmaceutical, medical device manufacturing and electronics industries
  • Suitable for use in cleanroom class ISO 6 and higher
  • Not clean laundered, not sterilized
TRUSTED_SUSTAINABILITY
Specifications
Size Large
Material Tyvek
Color White
Disposable Single-use
Closure Type Elastic Ankle