missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ U-251 Whole Cell Lysate

Product Code. 18333682 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18333682 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18333682 Supplier Novus Biologicals™ Supplier No. NBP306998
  • ONLINE EXCLUSIVE PRICE
    274.00€
    258.30€ / 500µg
    Save 15.70€
    5% Off
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

for use as a positive control in Western Blot

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) This product has been tested by SDS-PAGE and western blot. Ready-to-use lysates are especially prepared as positive controls for separation by SDS-PAGE and subsequent western blot analysis. Lysates are prepared in denaturing buffer WITHOUT dissociating agents (i.e. no 2-mercaptoethanol or dithiothreitol has been added). Heat lysate to 95C for 5 minutes and rapidly cool. If dissociating conditions are desired, add reducing agent prior to heating. The recommended loading volume per lane is 10-20 uL depending on the size format of your gel.
Quantity 500 μg
Purity Multi-step
Content And Storage Store at -70C. Avoid freeze-thaw cycles.
Target U-251
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.