Learn More
Invitrogen™ U2AF2 Recombinant Rabbit Monoclonal Antibody (24GB2465)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557810
 View more versions of this product
Product Code. 30274968
  • 564.00€ / 100µL
Description
U2AF2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
U2 auxiliary factor (U2AF), comprised of a large and a small subunit, is a non-snRNP protein required for the binding of U2 snRNP to the pre-mRNA branch site. This gene encodes the U2AF large subunit which contains a sequence-specific RNA-binding region with 3 RNA recognition motifs and an Arg/Ser-rich domain necessary for splicing. The large subunit binds to the polypyrimidine tract of introns early during spliceosome assembly. Multiple transcript variants have been detected for this gene, but the full-length natures of only two have been determined to date.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
U2AF2
Recombinant Monoclonal
Unconjugated
U2AF2
65kDa; hU2AF(65); hU2AF65; splicing factor U2AF 65 kDa subunit; U2 (RNU2) small nuclear RNA auxiliary factor 2; U2 auxiliary factor 65 kDa subunit; U2 small nuclear ribonucleoprotein auxiliary factor (65kD); U2 small nuclear ribonucleoprotein auxiliary factor (U2AF) 2; U2 small nuclear ribonucleoprotein auxiliary factor (U2AF), 65 kDa; U2 small nuclear RNA auxiliary factor 2; U2 snRNP auxiliary factor large subunit; U2AF2; U2af65; Unknown (protein for MGC:137461)
Rabbit
Affinity Chromatography
RUO
11338, 22185, 308335
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2465
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P26368, P26369
U2AF2
A synthesized peptide derived from human U2AF65 (aa 415-475).
100 μL
Primary
Human, Mouse, Rat