missing translation for 'onlineSavingsMsg'
Learn More
Learn More
U2AF35 Antibody (1E9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43059
This item is not returnable.
View return policy
Description
U2AF35 Monoclonal antibody specifically detects U2AF35 in Human samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| U2AF35 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 1000000000 | |
| Western Blot 1:500 - 1:5000, ELISA, Immunohistochemistry 1:20 - 1:200, Immunoprecipitation 1:200 - 1:2000 | |
| FP793, human U2 snRNP auxiliary factor small subunit10U2AF35DKFZp313J1712, RN, RNU2AF1,35-KD subunit, U2 auxiliary factor 35 kDa subunit, U2 small nuclear RNA auxiliary factor 1, U2 small nuclear RNA auxillary factor 1, U2 snRNP auxiliary factor small subunit | |
| U2 small nuclear RNA auxiliary factor 1 - (Uniprot# Q01081) | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 7307 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction