missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ UACA Antibody (UACA/1222), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27919
This item is not returnable.
View return policy
Description
UACA Monoclonal antibody specifically detects UACA in Human,Mouse samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| UACA | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry 0.5 - 1 μg/mL, Immunocytochemistry/ Immunofluorescence 0.5 - 1 μg/mL | |
| Q9BZF9 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| UACA/1222 | |
| Unconjugated | |
| PBS, 0.1 mg/mL BSA | |
| FLJ10128, KIAA1561MGC141969, MGC141967, nuclear membrane binding protein, NUCLING, uveal autoantigen with coiled coil domains and ankyrin repeats, uveal autoantigen with coiled-coil domains and ankyrin repeats | |
| Recombinant human protein was used as the immunogen for the UACA antibody. | |
| 100 μg | |
| Apoptosis | |
| 55075 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction