Learn More
UBA2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15347-1-AP-20UL
 View more versions of this product
Product Code. 16894484
  • 145.00€ / 20µL
Description
UBA2 is involved in covalent attachment of one protein to another is one of the more prominent posttranslational modifications in respect to size and ubiquity. Ubiquitin is the most familiar of the protein modifiers and its activation and transfer to target proteins has been studied extensively. Recently, a new group of ubiquitin-like proteins has been receiving a lot of attention. SUMO, or Sentrin, is one of the most intriguing. SUMO has been implicated in the stabilization of target proteins and/or their localization to sub-cellular complexes.The conjugation of SUMO-1, SUMO-2 and SUMO-3 onto target proteins requires the concerted action of the specific E1-activating enzyme SAE1/SAE2, the E2-conjugating enzyme Ubc9, and an E3-like SUMO ligase.
Specifications
Specifications
UBA2
Polyclonal
Unconjugated
UBA2
ARX, HRIHFB2115, SAE2, UBA2, UBLE1B
Rabbit
Antigen Affinity Chromatography
RUO
10054, 50995
-20°C
Liquid
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UBT2, Q9Z1F9
Uba2
UBA2 Fusion Protein Ag7586
20 μL
Primary
Human, Mouse