Learn More
SI Analytics™ Ubbelohde Viscometers for Manual Measurements
Product Code. 12058163 Shop All SI Analytics Products
Change view
Click to view available options
Viscosity Range:
0.8 to 5 cSt
50 to 500 cSt
Approximate Constant:
0.005
0.5
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Viscosity Range Approximate Constant Unit Size
12058163 50 to 500 cSt 0.5 Each
12008163 0.8 to 5 cSt 0.005 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12058163 Supplier SI Analytics™ Supplier No. 285400086
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
SI Analytics™ Ubbelohde Viscometers for Manual Measurements feature a suspended ball level for determination of absolute and relative kinematic viscosity of liquids with Newtonian flow behavior.
  • Calibrated, with constant, for manual measurement
  • Ideal for transparent liquids manual measurement, foaming liquids, liquid mixture with highly volatile components, and high-temperature or low temperature measurements
  • Delivered with manufacturer's certificate in accordance with DIN 55 350, Part 18
  • All viscometers are provided with ring marks
  • This ensures that viscometers for automatic measurements can also be checked by means of manual measurements
  • The recommended minimum flowthrough time is 200 seconds
TRUSTED_SUSTAINABILITY
Specifications
Product Type Ubbelohde Viscometer
Viscosity Range 50 to 500 cSt
Calibrated Yes
Description 1.62 mm diameter
Approximate Constant 0.5
Certifications/Compliance DIN 51 562 Part 1, ISO/DIS 3105 (BS-IP-SL)
Material Glass
Length (Metric) 290 mm
For Use With (Application) Manual measurements
Volume (Metric) 15 to 20 mL