Learn More
UBE2S Mouse anti-Human, Unconjugated, Clone: 8C9E6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66897-1-IG-150UL
 View more versions of this product
Product Code. 16839175
  • 410.00€ / 150µL
Description
The UBE2S antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UBE2S. This antibody recognizes human antigen. The UBE2S antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
UBE2S
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
8C9E6
Unconjugated
150 μL
27338
IgG2b