Learn More
UBE2S Mouse anti-Human, Unconjugated, Clone: 8C9E6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66897-1-IG-20UL
Description
The UBE2S antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UBE2S. This antibody recognizes human antigen. The UBE2S antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| UBE2S | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, ELISA | |
| 8C9E6 | |
| Unconjugated | |
| 20 μL | |
| 27338 | |
| IgG2b |