Learn More
UBE3B Monoclonal Antibody (OTI3F10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806725S
Description
UBE3B Monoclonal antibody specifically detects UBE3B in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| UBE3B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y6W6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 11221 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin) | |
| OTI3F10 | |
| Unconjugated | |
| DUSP10 | |
| BPIDS; HECT-type ubiquitin transferase E3B; KOS; UBE3B; UBE3B_v2; ubiquitin protein ligase E3B; ubiquitin-protein ligase E3B | |
| Human recombinant protein fragment corresponding to amino acids 180-482 of human DUSP10 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.