missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBOX5 Monoclonal Antibody (OTI 7G1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
242.00€ - 540.87€
Specifications
| Antigen | UBOX5 |
|---|---|
| Clone | OTI 7G1 |
| Concentration | 0.87 mg/mL |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17597811
|
Origene Technologies
TA502810S |
30 μL |
242.00€
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17577811
|
Origene Technologies
TA502810 |
100 μL |
540.87€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UBOX5 Monoclonal antibody specifically detects UBOX5 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| UBOX5 | |
| 0.87 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| O94941 | |
| IgG2a | |
| -20° C, Avoid Freeze/Thaw Cycles |
| OTI 7G1 | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| 22888 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title