missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBXN6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97298-50ul
This item is not returnable.
View return policy
Description
UBXN6 Polyclonal antibody specifically detects UBXN6 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| UBXN6 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| DKFZp667D109, FLJ00394, UBX domain containing 1, UBX domain protein 6, UBX domain-containing 1, UBX domain-containing 2, UBX domain-containing protein 1, UBX domain-containing protein 6, UBXD1, UBXDC2 | |
| Produced in rabbits immunized with E. coli-derived Human UBXN6 fragment. | |
| 50 μg | |
| Cell Biology | |
| 80700 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction