Learn More
UCH-L1/PGP9.5 Antibody (CL3210) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-46621MFV450
Product Code. 30299461
  • 759.00€ / 0.10mL
Description
UCH-L1/PGP9.5 Monoclonal antibody specifically detects UCH-L1/PGP9.5 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
UCH-L1/PGP9.5
Monoclonal
mFluor Violet 450 SE
EC 3.4.19.12, EC 6.-, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP9.5Uch-L1, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquitin thioesterase L1, UCHL1, UCH-L1
This antibody was developed using a recombinant protein derived from P09936, with the exact immunogen sequence remaining proprietary.
0.1 mL
Alzheimers Research, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission
7345
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL3210
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat