Learn More
UEVLD Monoclonal Antibody (OTI11E2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809207
Encompass_Preferred
 View more versions of this product
Product Code. 17581204
  • 540.87€ / 100µL
Description
Possible negative regulator of polyubiquitination.
Specifications
Specifications
UEVLD
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8IX04
UEVLD
Human recombit protein fragment corresponding to amino acids 131-379 of human UEVLD produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI11E2
Unconjugated
UEVLD
ATTP, UEV3
Mouse
Affinity Chromatography
RUO
55293
-20° C, Avoid Freeze/Thaw Cycles