Learn More
Fisherbrand™ UK Signal Atomic Clock
Product Code. 15368754 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15368754 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 15368754
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Use this clock for general laboratory time keeping applications. Fisherbrand™ Traceable™ UK Signal Atomic Clock automatically sets the correct date and time.
  • Features: radio signal atomic clock automatically sets correct date/time; calendar displays month/date and day
  • Temperature range: -5°C to 50°C (23°F to 122°F); resolution: 0.1°C; accuracy: ±1°C
  • Supplied with wall mount, bench stand, batteries, Traceable™ Certificate
Order Info
DisplayType
Specifications
Product Type UK Signal Atomic Clock
Accuracy ±1°C
Temperature Range (Metric) -5°C to +50°C
Temperature Accuracy ±1°C
Temperature Resolution 0.1°C
Quantity 1 Each
Product Title
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.