missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UL27 Rabbit anti-Virus, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10363-1-AP-150UL
This item is not returnable.
View return policy
Description
Promotes a cell cycle arrest in G0/G1 by inducing the proteasomal degradation of host histone acetyltransferase KAT5/Tip60.Specifications
| UL27 | |
| Polyclonal | |
| Unconjugated | |
| UL27 | |
| UL27 Fusion Protein Ag0411 | |
| 150 μL | |
| Primary | |
| -20°C | |
| Liquid |
| ELISA | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction