missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULK1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-41217-0.025mg
This item is not returnable.
View return policy
Description
ULK1 Polyclonal specifically detects ULK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| ULK1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| ULK1 | |
| Antibody was raised against a 16 amino acid peptide near the center of human ULK1 . The immunogen is located within amino acids 350 - 400 of ULK1. Amino Acid Squence: CDLVAEAPSAKPPPDSL | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Two alternatively spliced transcript variants encoding different isoforms have been identified. ULK1 antibody is predicted to not cross-react with ULK2. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500, Immunofluorescence | |
| ATG1, ATG1 autophagy related 1 homolog, ATG1A, EC 2.7.11, EC 2.7.11.1, FLJ38455, FLJ46475, KIAA0722, serine/threonine-protein kinase ULK1, UNC51, unc-51 (C. elegans)-like kinase 1, Unc51.1, Unc-51-like kinase 1, unc-51-like kinase 1 (C. elegans) | |
| Rabbit | |
| 105 kDa | |
| 0.025 mg | |
| Autophagy, mTOR Pathway, Protein Kinase | |
| 8408 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction